This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FILIP1L SIRNA Assays ›
Gene Symbol:
FILIP1L
Gene Name:
filamin A interacting protein 1 like
Gene Aliases:
DOC-1, DOC1, GIP130, GIP90
Chromosome Location:
Chr.3: 99833144 - 100114513 on Build GRCh38
UniGene ID:
Hs.104672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126552

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FILIP1L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042459.2 NP_001035924.1 5 3374
NM_001282793.1 NP_001269722.1 3 1925
NM_001282794.1 NP_001269723.1 2 2430
NM_014890.3 NP_055705.2 2 2430
NM_182909.3 NP_878913.2 5 3374
XM_006713486.2 XP_006713549.1 3 2626
XM_011512371.2 XP_011510673.1 3 2492
GenBank mRNA AF329092.1 - 5 3376
AF514867.1 - 4 2903
AF514868.1 - 4 2903
AF514869.1 - 4 2903
AK096195.1 - 3 2225
AK225855.1 - 3 2571
AX776292.1 - 5 3376
AX776294.1 - 4 2911
AX776296.1 - 4 2914
AX776298.1 - 4 2911
AY642382.1 - 4 2918
BC027860.1 - 2 2361 5228947
EU531865.1 - 2 2332
EU531866.1 - 3 1831
U53445.1 - 2 2275

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0