This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CATSPERD SIRNA Assays ›
Gene Symbol:
CATSPERD
Gene Name:
cation channel sperm associated auxiliary subunit delta
Gene Aliases:
TMEM146
Chromosome Location:
Chr.19: 5720385 - 5778734 on Build GRCh38
UniGene ID:
Hs.631842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126199

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CATSPERD
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152784.3 NP_689997.3 12, 13 1218
XM_011527885.2 XP_011526187.1 12, 13 1510
XM_011527886.2 XP_011526188.1 12, 13 1456
XM_011527887.2 XP_011526189.1 12, 13 1510
XM_011527888.2 XP_011526190.1 12, 13 1510
XM_011527889.2 XP_011526191.1 12, 13 1510
XM_011527890.2 XP_011526192.1 12, 13 1510
XM_011527891.1 XP_011526193.1 3, 4 306
XM_017026564.1 XP_016882053.1 11, 12 1426
XM_017026565.1 XP_016882054.1 11, 12 1163
XM_017026566.1 XP_016882055.1 8, 9 1096
GenBank mRNA AK128088.1 - 12, 13 1226
BC043005.1 - 12, 13 1204 5297674

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
sperm motility
response to progesterone
sperm-egg recognition
sperm capacitation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0