This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WBP2NL SIRNA Assays ›
Gene Symbol:
WBP2NL
Gene Name:
WBP2 N-terminal like
Gene Aliases:
GRAMD7, PAWP
Chromosome Location:
Chr.22: 41998725 - 42058456 on Build GRCh38
UniGene ID:
Hs.655541
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126163

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: WBP2NL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152613.2 NP_689826.2 6 2084
GenBank mRNA BC022546.1 - 6 2021 4811804
BC022549.1 - 6 2020 4815849

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

egg activation
female pronucleus assembly
male pronucleus assembly
meiotic cell cycle
WW domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm