This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01106 SIRNA Assays ›
Gene Symbol:
LINC01106 LINC01123
Gene Name:
long intergenic non-protein coding RNA 1106
long intergenic non-protein coding RNA 1123
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 110375109 - 110384536 on Build GRCh38
Chromosome Location:
Chr.2: 109986980 - 109995281 on Build GRCh38
UniGene ID:
Hs.643369
Hs.516403
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 126127

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01106
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027244.1 2 974
GenBank mRNA AK095678.1 - 2 1283
CR749351.1 - 3 1065
Gene Symbol: LINC01123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046110.1 4 2390
NR_046111.1 3 2205
NR_046112.1 2 963
GenBank mRNA AK056084.1 - 4 2307

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header