This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ERICH6 SIRNA Assays ›
Gene Symbol:
ERICH6
Gene Name:
glutamate rich 6
Gene Aliases:
C3orf44, ERICH6A, FAM194A
Chromosome Location:
Chr.3: 150659885 - 150704563 on Build GRCh38
UniGene ID:
Hs.147128
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126093

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ERICH6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308234.1 NP_001295163.1 7, 8 648
NM_152394.4 NP_689607.2 7, 8 952
XM_005247120.3 XP_005247177.1 7, 8 1544
GenBank mRNA AK093375.1 - 8, 9 787
AL834214.1 - 452
BC029577.1 - 7, 8 632 5268183
BC036239.1 - 7, 8 936 5297417

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0