This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM128 SIRNA Assays ›
Gene Symbol:
TMEM128
Gene Name:
transmembrane protein 128
Gene Aliases:
-
Chromosome Location:
Chr.4: 4235542 - 4248265 on Build GRCh38
UniGene ID:
Hs.12845
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125783

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM128
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297551.1 NP_001284480.1 3 400
NM_001297552.1 NP_001284481.1 3 400
NM_032927.3 NP_116316.1 3 905
XM_005248034.2 XP_005248091.1 3 423
GenBank mRNA AK295248.1 - 3 382
AK296007.1 - 3 381
BC007729.2 - 3 870 4303698
BC091482.1 - 3 400 30344390
BM263799.1 - 3 382
CR933611.1 - 3 390
DA019506.1 - 3 390

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0