This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF671 SIRNA Assays ›
Gene Symbol:
ZNF671
Gene Name:
zinc finger protein 671
Gene Aliases:
-
Chromosome Location:
Chr.19: 57719747 - 57727637 on Build GRCh38
UniGene ID:
Hs.180402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125543

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF671
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321375.1 NP_001308304.1 3 626
NM_001321376.1 NP_001308305.1 5 791
NM_024833.2 NP_079109.2 4 756
XM_017027314.1 XP_016882803.1 4 1249
GenBank mRNA AK027159.1 - 4 744
AK091421.1 - 5 791
AK225665.1 - 4 753
AK293680.1 - 3 624
BC025728.1 - 4 703 5202908

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
DNA binding
transcription cofactor activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0