This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM124B SIRNA Assays ›
Gene Symbol:
FAM124B
Gene Name:
family with sequence similarity 124 member B
Gene Aliases:
-
Chromosome Location:
Chr.2: 224378698 - 224401994 on Build GRCh38
UniGene ID:
Hs.147585
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125536

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM124B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122779.1 NP_001116251.1 2 1509
NM_024785.2 NP_079061.2 3 1634
GenBank mRNA AK075126.1 - 3 1137
BC025754.1 - 3 1634 5202295

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0