This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FASTKD1 SIRNA Assays ›
Gene Symbol:
FASTKD1
Gene Name:
FAST kinase domains 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 169527879 - 169573921 on Build GRCh38
UniGene ID:
Hs.529276
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125480

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FASTKD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281476.2 NP_001268405.1 12 2511
NM_001322046.1 NP_001308975.1 13 3313
NM_001322048.1 NP_001308977.1 12 2568
NM_001322049.1 NP_001308978.1 11 2442
NM_024622.5 NP_078898.3 13 2640
NR_104020.2 12 2507
XM_006712750.3 XP_006712813.1 12 2566
XM_006712751.3 XP_006712814.1 9 2651
XM_017004911.1 XP_016860400.1 13 3953
XM_017004912.1 XP_016860401.1 8 2522
GenBank mRNA AB058703.1 - 11 1822
AK025554.1 - 10 1752
AK055892.1 - 12 2422
AK074302.1 - 13 2531
AK130797.1 - 12 2431
AK225293.1 - 10 1752
AL832058.1 - 13 2523
BC032687.2 - 12 1969 5581443

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cellular respiration
protein kinase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0