This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FASTKD5 SIRNA Assays ›
Gene Symbol:
FASTKD5
Gene Name:
FAST kinase domains 5
Gene Aliases:
dJ1187M17.5
Chromosome Location:
Chr.20: 3146519 - 3159886 on Build GRCh38
UniGene ID:
Hs.741168
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125331

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FASTKD5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021826.4 NP_068598.1 2 2644
GenBank mRNA AB058695.1 - 1 5010
AK023211.1 - 2 2623
AK027215.1 - 1 1127
AK301748.1 - 3 2394
BC007413.2 - 2 2644 3010170
BC011856.1 - 2 2608 4298806

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial RNA processing
mRNA processing
protein phosphorylation
cellular respiration
protein kinase activity
protein binding
rRNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0