This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIAA1524 SIRNA Assays ›
Gene Symbol:
KIAA1524
Gene Name:
KIAA1524
Gene Aliases:
CIP2A, p90
Chromosome Location:
Chr.3: 108545752 - 108589644 on Build GRCh38
UniGene ID:
Hs.591308
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125308

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA1524
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020890.2 NP_065941.2 3 609
XM_006713716.3 XP_006713779.1 3 393
XM_006713717.2 XP_006713780.1 4 505
XM_011513056.1 XP_011511358.1 3 417
XM_011513057.2 XP_011511359.1 4 505
GenBank mRNA AB040957.1 - 3 336
AF334474.1 - 3 408
AK056933.1 - 3 418
AK308315.1 - 3 419
AK310446.1 - 3 613
AL713689.1 - 3 404
BC063433.1 - 3 400
BC082961.1 - 3 362
BC130564.1 - 3 376 40146595
BC136371.1 - 3 376 9020358

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
cell-cell adhesion
positive regulation of neural precursor cell proliferation
protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0