This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SF3B6 SIRNA Assays ›
Gene Symbol:
SF3B6
Gene Name:
splicing factor 3b subunit 6
Gene Aliases:
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a
Chromosome Location:
Chr.2: 24067584 - 24076444 on Build GRCh38
UniGene ID:
Hs.177861
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 124936

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SF3B6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016047.3 NP_057131.1 3 451
GenBank mRNA AF151868.1 - 3 447
AF161523.1 - 3 345
AF184213.1 - 3 447
AF401310.1 - 3 236
AK000621.1 - 3 351
AK311118.1 - 2 238
AK311908.1 - 3 384
BC015463.1 - 3 451 3914184
BG200650.1 - 2 212

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
blastocyst formation
nucleotide binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0