This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AKNAD1 SIRNA Assays ›
Gene Symbol:
AKNAD1
Gene Name:
AKNA domain containing 1
Gene Aliases:
C1orf62
Chromosome Location:
Chr.1: 108815898 - 108858242 on Build GRCh38
UniGene ID:
Hs.729441
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 124124

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AKNAD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152763.4 NP_689976.2 2 860
NR_049760.1 3 1001
GenBank mRNA AK095517.1 - 3 987
AK313772.1 - 2 840
AL832216.1 - 2 850
BC030647.2 - 2 840
BC150642.1 - 1 615 9057012

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0