This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC8C SIRNA Assays ›
Gene Symbol:
LRRC8C
Gene Name:
leucine rich repeat containing 8 family member C
Gene Aliases:
AD158, FAD158
Chromosome Location:
Chr.1: 89615810 - 89719535 on Build GRCh38
UniGene ID:
Hs.412836
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 123658

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC8C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032270.4 NP_115646.2 3 1456
XM_006710960.3 XP_006711023.1 3 3098
XM_011542282.2 XP_011540584.1 3 1443
XM_011542283.2 XP_011540585.1 4 3010
XM_017002503.1 XP_016857992.1 3 1303
XM_017002504.1 XP_016857993.1 4 1212
GenBank mRNA AB081509.1 - 3 1337
AK127881.1 - 3 1393
AL136919.1 - 1 956
BC113973.1 - 3 1263 40007964

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of anion transport
fat cell differentiation
anion transmembrane transport
volume-sensitive anion channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0