This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIAA0319L SIRNA Assays ›
Gene Symbol:
KIAA0319L
Gene Name:
KIAA0319 like
Gene Aliases:
AAVR
Chromosome Location:
Chr.1: 35433490 - 35557670 on Build GRCh38
UniGene ID:
Hs.456507
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 123515

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA0319L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024874.4 NP_079150.3 3, 4 932
XM_006710907.1 XP_006710970.1 3, 4 731
XM_006710909.1 XP_006710972.1 3, 4 823
XM_011542179.2 XP_011540481.1 3, 4 1063
XM_011542180.1 XP_011540482.1 3, 4 901
XM_017002366.1 XP_016857855.1 3, 4 901
XM_017002367.1 XP_016857856.1 3, 4 1063
XM_017002368.1 XP_016857857.1 3, 4 731
XM_017002369.1 XP_016857858.1 3, 4 823
XM_017002370.1 XP_016857859.1 3, 4 901
XM_017002371.1 XP_016857860.1 3, 4 901
XM_017002372.1 XP_016857861.1 3, 4 823
XM_017002373.1 XP_016857862.1 3, 4 901
GenBank mRNA AF275679.2 - 3, 4 881
AF289597.1 - 1, 2 497
AK302428.1 - 3, 4 942
AK314605.1 - 3, 4 880
AK316252.1 - 3, 4 1060
AY163234.1 - 3, 4 881
BC014530.1 - 3, 4 864 3843301

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0