This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other QPRT SIRNA Assays ›
Gene Symbol:
QPRT
Gene Name:
quinolinate phosphoribosyltransferase
Gene Aliases:
HEL-S-90n, QPRTase
Chromosome Location:
Chr.16: 29670588 - 29698699 on Build GRCh38
UniGene ID:
Hs.513484
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 122453

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: QPRT
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318249.1 NP_001305178.1 3 824
NM_001318250.1 NP_001305179.1 4 916
NM_014298.4 NP_055113.2 4 1360
NR_134536.1 3 853
XM_005255223.3 XP_005255280.2 5 1446
XM_017023101.1 XP_016878590.1 5 1443
GenBank mRNA AK090801.1 - 1 1895
AK293195.1 - 3 824
BC005060.1 - 4 1197 2960170
BC010033.2 - 4 1189 3506460
EU794638.1 - 4 1189

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

NAD biosynthetic process
NAD metabolic process
quinolinate catabolic process
protein oligomerization
nicotinate-nucleotide diphosphorylase (carboxylating) activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0