This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IGFBP7 SIRNA Assays ›
Gene Symbol:
IGFBP7
Gene Name:
insulin like growth factor binding protein 7
Gene Aliases:
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF
Chromosome Location:
Chr.4: 57031071 - 57110385 on Build GRCh38
UniGene ID:
Hs.479808
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 122165

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IGFBP7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253835.1 NP_001240764.1 4 783
NM_001553.2 NP_001544.1 4 783
GenBank mRNA AK303915.1 - 4 785
AK316082.1 - 4 783
BC017201.2 - 4 771 3632247
BC066339.1 - 4 783 5296697
BT006654.1 - 4 749
L19182.1 - 4 762
S75725.1 - 4 771

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
cell adhesion
negative regulation of cell proliferation
protein binding
insulin-like growth factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0