This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OSBPL10 SIRNA Assays ›
Gene Symbol:
OSBPL10
Gene Name:
oxysterol binding protein like 10
Gene Aliases:
ORP10, OSBP9
Chromosome Location:
Chr.3: 31660825 - 31981850 on Build GRCh38
UniGene ID:
Hs.150122
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 121043
Validated

Purification
Size
Availability Inventoried
Catalog # AM51331
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OSBPL10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174060.1 NP_001167531.1 6 1669
NM_017784.4 NP_060254.2 7 1861
XM_005264843.3 XP_005264900.1 7 1637
XM_005264844.4 XP_005264901.1 6 1445
XM_005264845.1 XP_005264902.1 2 545
XM_011533326.2 XP_011531628.1 7 1014
XM_017005667.1 XP_016861156.1 7 1527
XM_017005668.1 XP_016861157.1 7 1504
XM_017005669.1 XP_016861158.1 7 1033
XM_017005670.1 XP_016861159.1 4 1716
GenBank mRNA AB209872.1 - 4 496
AF392451.1 - 8 1571
AK000370.1 - 5 737
AK304067.1 - 6 1122
AL833247.1 - 7 964
BC003168.1 - 7 1265 2905981

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00215016_m1

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid transport
phosphatidylserine binding
cholesterol binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0