This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TEN1-CDK3 SIRNA Assays ›
Gene Symbol:
TEN1-CDK3 CDK3
Gene Name:
TEN1-CDK3 readthrough (NMD candidate)
cyclin dependent kinase 3
Gene Aliases:
C17orf106-CDK3
-
Chromosome Location:
Chr.17: 75979217 - 76005999 on Build GRCh38
Chromosome Location:
Chr.17: 76000906 - 76005999 on Build GRCh38
UniGene ID:
Hs.706766
Hs.706766
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 119438

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TEN1-CDK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037709.1 5 1981
GenBank mRNA AK097104.1 - 5 1938
BX647274.1 - 5 1967
Gene Symbol: CDK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258.2 NP_001249.1 3 221
GenBank mRNA DB455174.1 - 3 222
X66357.1 - 2 233

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein phosphorylation
cellular response to DNA damage stimulus
mitotic nuclear division
cell proliferation
G0 to G1 transition
cell division
regulation of cell cycle
cyclin-dependent protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0