This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCNB3 SIRNA Assays ›
Gene Symbol:
CCNB3
Gene Name:
cyclin B3
Gene Aliases:
-
Chromosome Location:
Chr.X: 50202713 - 50351914 on Build GRCh38
UniGene ID:
Hs.130310
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 118848

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCNB3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033031.2 NP_149020.2 9 3842
NM_033670.2 NP_391990.1 5 530
XM_017029913.1 XP_016885402.1 11 4053
XM_017029914.1 XP_016885403.1 11 3983
XM_017029915.1 XP_016885404.1 12 4013
XM_017029916.1 XP_016885405.1 11 4050
XM_017029917.1 XP_016885406.1 10 4029
XM_017029918.1 XP_016885407.1 10 4026
GenBank mRNA AJ314764.1 - 9 3846
AJ314765.1 - 5 534
AJ314766.1 - 8 854
AJ416458.1 - 9 3858
AK292454.1 - 10 4015
AY927611.1 - 2 707

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of G2/M transition of mitotic cell cycle
cell division
meiotic cell cycle
protein binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0