This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEACAM6 SIRNA Assays ›
Gene Symbol:
CEACAM6
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 6
Gene Aliases:
CD66c, CEAL, NCA
Chromosome Location:
Chr.19: 41755421 - 41772211 on Build GRCh38
UniGene ID:
Hs.466814
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 11706

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CEACAM6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002483.6 NP_002474.4 3 747
XM_011526990.1 XP_011525292.1 3 846
GenBank mRNA AK312542.1 - 3 732
BC005008.1 - 3 668 3640231
BT009774.1 - 3 628
M18216.1 - 3 674
M18728.1 - 3 678
M29541.1 - 3 721

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell-cell signaling
leukocyte migration
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0