This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF169 SIRNA Assays ›
Gene Symbol:
ZNF169
Gene Name:
zinc finger protein 169
Gene Aliases:
-
Chromosome Location:
Chr.9: 94259266 - 94303967 on Build GRCh38
UniGene ID:
Hs.387623
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116770

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF169
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301275.1 NP_001288204.1 5 907
NM_003448.2 NP_003439.2 5 543
NM_194320.3 NP_919301.2 5 540
XM_017014362.1 XP_016869851.1 5 672
XM_017014363.1 XP_016869852.1 6 566
XM_017014364.1 XP_016869853.1 6 563
XM_017014365.1 XP_016869854.1 5 6896
XM_017014366.1 XP_016869855.1 5 6834
GenBank mRNA AK122832.1 - 1 1163
AK289742.1 - 5 484
BC047702.2 - 4 464 5585128
BP873364.1 - 5 495
DA490480.1 - 5 511

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0