This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF166 SIRNA Assays ›
Gene Symbol:
RNF166
Gene Name:
ring finger protein 166
Gene Aliases:
-
Chromosome Location:
Chr.16: 88696495 - 88706421 on Build GRCh38
UniGene ID:
Hs.513804
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116732

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF166
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171816.1 NP_001165287.1 2 948
NM_178841.3 NP_849163.1 2 371
XM_011522845.2 XP_011521147.1 2 365
XM_011522846.1 XP_011521148.1 2 371
XM_017022910.1 XP_016878399.1 1 2956
XM_017022911.1 XP_016878400.1 2 365
XM_017022912.1 XP_016878401.1 2 365
XM_017022913.1 XP_016878402.1 2 1602
GenBank mRNA AK057106.1 - 2 948
BC013948.2 - 2 297 3509621
BC017226.1 - 2 349 4299817
DA376321.1 - 2 371

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
zinc ion binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm