This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF329 SIRNA Assays ›
Gene Symbol:
ZNF329
Gene Name:
zinc finger protein 329
Gene Aliases:
-
Chromosome Location:
Chr.19: 58126328 - 58155169 on Build GRCh38
UniGene ID:
Hs.458377
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116372

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF329
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024620.3 NP_078896.3 4 805
XM_006723381.2 XP_006723444.1 3 742
XM_006723382.3 XP_006723445.1 2 751
XM_006723383.3 XP_006723446.1 2 1256
XM_006723384.3 XP_006723447.1 2 955
XM_011527310.2 XP_011525612.1 5 1189
XM_011527311.2 XP_011525613.1 4 1015
XM_011527312.2 XP_011525614.1 4 780
XM_011527313.2 XP_011525615.1 4 1092
XM_011527314.2 XP_011525616.1 4 1071
XM_011527315.1 XP_011525617.1 4 1151
XM_011527316.2 XP_011525618.1 2 631
XM_017027307.1 XP_016882796.1 4 1455
XM_017027308.1 XP_016882797.1 3 1361
XM_017027309.1 XP_016882798.1 3 1116
XM_017027310.1 XP_016882799.1 3 1417
XM_017027311.1 XP_016882800.1 2 928
GenBank mRNA AK022648.1 - 1 98
AK090893.1 - 4 1151
AK095349.1 - 5 1246
AK096542.1 - 2 612
BC047501.1 - 4 802 5266873

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0