This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF574 SIRNA Assays ›
Gene Symbol:
ZNF574
Gene Name:
zinc finger protein 574
Gene Aliases:
FP972
Chromosome Location:
Chr.19: 42068477 - 42081567 on Build GRCh38
UniGene ID:
Hs.13323
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116348

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF574
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022752.5 NP_073589.4 2 246
XM_005259159.2 XP_005259216.1 1 109
XM_011527228.2 XP_011525530.1 2 439
XM_011527229.2 XP_011525531.1 2 162
XM_011527230.2 XP_011525532.1 2 114
XM_017027149.1 XP_016882638.1 2 582
GenBank mRNA AF447880.1 - 2 190
AK027895.1 - 2 149
AK131369.1 - 2 582
AK131418.1 - 2 732
BC001184.1 - 2 135 3502564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0