This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF124 SIRNA Assays ›
Gene Symbol:
ZNF124
Gene Name:
zinc finger protein 124
Gene Aliases:
HZF-16, HZF16, ZK7
Chromosome Location:
Chr.1: 247121975 - 247172378 on Build GRCh38
UniGene ID:
Hs.421238
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115407

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF124
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297568.1 NP_001284497.1 4 620
NM_001297569.1 NP_001284498.1 4 424
NM_003431.3 NP_003422.2 4 434
XM_005273257.4 XP_005273314.1 4 487
XM_006711813.3 XP_006711876.1 4 673
XM_011544272.2 XP_011542574.1 4 471
GenBank mRNA AK054647.1 - 4 433
BC096349.4 - 4 384 40009545
BC096350.4 - 4 384 40009546
BC096351.2 - 4 570 40009547
BC099661.3 - 4 570 40009541
DA911997.1 - 4 434
DN993107.1 - 4 410
S54641.1 - 1 825

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0