This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF84 SIRNA Assays ›
Gene Symbol:
ZNF84
Gene Name:
zinc finger protein 84
Gene Aliases:
HPF2
Chromosome Location:
Chr.12: 133037286 - 133063304 on Build GRCh38
UniGene ID:
Hs.445019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115405

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF84
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127372.2 NP_001120844.1 5 1030
NM_001289971.1 NP_001276900.1 5 1021
NM_001289972.1 NP_001276901.1 5 1149
NM_003428.5 NP_003419.3 5 1014
XM_005266185.1 XP_005266242.1 5 1184
XM_005266186.1 XP_005266243.1 6 920
XM_011534832.1 XP_011533134.1 5 964
GenBank mRNA AK314232.1 - 5 792
BC036656.1 - 5 1136 4828620
BC048350.1 - 4 598 5555056
BC062552.1 - 5 790 6536224
M27878.1 - 5 922
X60156.2 - 4 760

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0