This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP2R5D SIRNA Assays ›
Gene Symbol:
PPP2R5D
Gene Name:
protein phosphatase 2 regulatory subunit B'delta
Gene Aliases:
B56D, MRD35
Chromosome Location:
Chr.6: 42984499 - 43012345 on Build GRCh38
UniGene ID:
Hs.533308
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 114634

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPP2R5D
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270476.1 NP_001257405.1 4 674
NM_006245.3 NP_006236.1 4 657
NM_180976.2 NP_851307.1 4 561
NM_180977.2 NP_851308.1 2 339
GenBank mRNA AB000634.1 - 4 492
AB000635.1 - 2 186
AB209861.1 - 2 236
AB451342.1 - 4 478
AB451357.1 - 4 478
AK122701.1 - 4 551
AK290604.1 - 4 470
AK299696.1 - 4 530
BC001095.2 - 4 541 3507280
BC001175.1 - 4 468 3355395
BC010692.1 - 4 551 3853892
D78360.1 - 4 459
L76702.1 - 4 665

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
signal transduction
nervous system development
regulation of protein phosphatase type 2A activity
protein serine/threonine phosphatase activity
protein binding
protein phosphatase type 2A regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0