This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACHD1 SIRNA Assays ›
Gene Symbol:
CACHD1
Gene Name:
cache domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 64470793 - 64693058 on Build GRCh38
UniGene ID:
Hs.443891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 113628

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CACHD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293274.1 NP_001280203.1 9 1653
NM_020925.2 NP_065976.2 10 1374
XM_011541862.1 XP_011540164.1 9 1649
XM_011541863.1 XP_011540165.1 10 1798
GenBank mRNA AB046793.1 - 8 1156
BC039301.1 - 10 1360 4836806
BX537603.1 - 9 1652

Back To Top

More Information


Gene Ontology Categories:

Function(s)

calcium ion transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0