This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSTO1 SIRNA Assays ›
Gene Symbol:
GSTO1
Gene Name:
glutathione S-transferase omega 1
Gene Aliases:
GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
Chromosome Location:
Chr.10: 104254194 - 104267464 on Build GRCh38
UniGene ID:
Hs.190028
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 111517

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSTO1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191002.1 NP_001177931.1 5 870
NM_001191003.1 NP_001177932.1 6 1022
NM_004832.2 NP_004823.1 6 969
GenBank mRNA AF212303.1 - 6 776
BC000127.1 - 6 779 3351359
BX537431.1 - 6 897
CN283947.1 - 5 673
EU794619.1 - 6 776
U90313.1 - 6 759

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
L-ascorbic acid metabolic process
L-ascorbic acid biosynthetic process
methylation
xenobiotic catabolic process
oxidation-reduction process
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0