This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZSCAN31 SIRNA Assays ›
Gene Symbol:
ZSCAN31
Gene Name:
zinc finger and SCAN domain containing 31
Gene Aliases:
ZNF20-Lp, ZNF310P, ZNF323
Chromosome Location:
Chr.6: 28324737 - 28358202 on Build GRCh38
UniGene ID:
Hs.656413
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 109674

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZSCAN31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135215.1 NP_001128687.1 8 3288
NM_001135216.1 NP_001128688.1 4 2784
NM_001243241.1 NP_001230170.1 4 2709
NM_001243242.1 NP_001230171.1 4 2406
NM_001243243.1 NP_001230172.1 3 2308
NM_001243244.1 NP_001230173.1 3 2297
NM_030899.4 NP_112161.3 4 2773
NM_145909.2 NP_665916.1 8 2910
XM_005249295.1 XP_005249352.1 4 2635
XM_005249296.4 XP_005249353.1 5 6910
XM_011514807.2 XP_011513109.1 6 6960
XM_011514808.2 XP_011513110.1 4 6756
XM_011514809.1 XP_011513111.1 4 2620
XM_011514811.2 XP_011513113.1 4 3135
XM_011514812.2 XP_011513114.1 4 2661
XM_011514813.2 XP_011513115.1 4 3124
XM_017011196.1 XP_016866685.1 6 7048
GenBank mRNA AF513019.1 - 8 2910
AK095261.1 - 8 3288
AL833416.1 - 3 2068
AL833617.1 - 4 2785
BC008490.1 - 4 2547 4248596
BC065241.1 - 3 2078 6016003

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0