This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LANCL2 SIRNA Assays ›
Gene Symbol:
LANCL2
Gene Name:
LanC like 2
Gene Aliases:
GPR69B, TASP
Chromosome Location:
Chr.7: 55365448 - 55433742 on Build GRCh38
UniGene ID:
Hs.595384
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 109155

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LANCL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018697.3 NP_061167.1 4 1190
GenBank mRNA AB035966.1 - 4 667
AF353942.1 - 4 667
AJ278245.1 - 4 798
AK095857.1 - 4 1190
AK225958.1 - 4 643
AK301747.1 - 4 596
AK313326.1 - 4 935
AL713665.2 - 4 582
BC004887.2 - 1 18
BC070049.1 - 4 886 30343188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of abscisic acid-activated signaling pathway
negative regulation of transcription, DNA-templated
catalytic activity
protein binding
ATP binding
GTP binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-4-phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0