This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD160 SIRNA Assays ›
Gene Symbol:
CD160
Gene Name:
CD160 molecule
Gene Aliases:
BY55, NK1, NK28
Chromosome Location:
Chr.1: 145719312 - 145739288 on Build GRCh38
UniGene ID:
Hs.488237
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 108175

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007053.3 NP_008984.1 4 542
XM_005272929.3 XP_005272986.1 3 570
XM_011509104.2 XP_011507406.1 4 676
GenBank mRNA AF060981.1 - 4 465
BC014465.1 - 4 468 4849145
CR541867.1 - 2 250
CR541888.1 - 2 250
EU016100.1 - 2 250

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular defense response
cell surface receptor signaling pathway
cell proliferation
regulation of immune response
defense response to Gram-negative bacterium
receptor activity
receptor binding
protein binding
MHC class I receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0