This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALDH4A1 SIRNA Assays ›
Gene Symbol:
ALDH4A1
Gene Name:
aldehyde dehydrogenase 4 family member A1
Gene Aliases:
ALDH4, P5CD, P5CDh
Chromosome Location:
Chr.1: 18871430 - 18902799 on Build GRCh38
UniGene ID:
Hs.77448
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 107234

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ALDH4A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161504.1 NP_001154976.1 10 1254
NM_001319218.1 NP_001306147.1 10 1394
NM_003748.3 NP_003739.2 10 1394
NM_170726.2 NP_733844.1 10 1394
GenBank mRNA AK289972.1 - 10 1320
AK294552.1 - 10 1254
BC007581.1 - 10 1124 3139944
BC023600.2 - 10 1135 4548787
FJ462711.1 - 10 1132
U24266.1 - 10 1148
U24267.1 - 10 1148

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glutamate biosynthetic process
proline metabolic process
proline biosynthetic process
proline catabolic process
proline catabolic process to glutamate
4-hydroxyproline catabolic process
oxidation-reduction process
1-pyrroline-5-carboxylate dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
electron carrier activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0