This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF135 SIRNA Assays ›
Gene Symbol:
ZNF135
Gene Name:
zinc finger protein 135
Gene Aliases:
ZNF61, ZNF78L1, pHZ-17, pT3
Chromosome Location:
Chr.19: 58059227 - 58069755 on Build GRCh38
UniGene ID:
Hs.85863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 107123

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF135
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164529.1 NP_001158001.1 4 2352
NM_001164530.1 NP_001158002.1 5 1871
NM_001289401.1 NP_001276330.1 5 2778
NM_001289402.1 NP_001276331.1 5 3141
NM_003436.3 NP_003427.3 5 2809
NM_007134.1 NP_009065.1 4 2747
XM_005259211.1 XP_005259268.1 4 2711
XM_006723362.3 XP_006723425.1 5 2817
XM_006723363.3 XP_006723426.1 5 3181
XM_017027240.1 XP_016882729.1 4 3134
XM_017027241.1 XP_016882730.1 5 3136
XM_017027242.1 XP_016882731.1 4 2576
GenBank mRNA AK094202.1 - 5 3100
AK098011.1 - 5 1871
AK297226.1 - 5 2425
AL833637.1 - 4 2310
BC046434.1 - 5 2773 4824328
BX647775.1 - 4 2715
U09413.1 - 1 2172

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header