This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC5L SIRNA Assays ›
Gene Symbol:
CDC5L
Gene Name:
cell division cycle 5 like
Gene Aliases:
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
Chromosome Location:
Chr.6: 44387514 - 44450424 on Build GRCh38
UniGene ID:
Hs.485471
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 106420

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDC5L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253.3 NP_001244.1 6 1005
GenBank mRNA AB007892.2 - 6 777
AK128737.1 - 6 813
AK299738.1 - 6 732
BC001568.1 - 6 794 3452352
U86753.1 - 6 695

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
mRNA splicing, via spliceosome
DNA repair
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell differentiation
cellular response to fibroblast growth factor stimulus
cellular response to interleukin-2
signal transduction involved in DNA damage checkpoint
cellular response to wortmannin
cellular response to nerve growth factor stimulus
cellular response to prolactin
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription corepressor binding
DNA binding
protein binding
protein phosphatase 1 binding
protein kinase binding
leucine zipper domain binding
sequence-specific DNA binding
transcription regulatory region DNA binding
poly(A) RNA binding
WD40-repeat domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header