This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC3 SIRNA Assays ›
Gene Symbol:
ZDHHC3
Gene Name:
zinc finger DHHC-type containing 3
Gene Aliases:
DHHC-3, GODZ, ZNF373
Chromosome Location:
Chr.3: 44915261 - 44976190 on Build GRCh38
UniGene ID:
Hs.61430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 105728

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZDHHC3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135179.1 NP_001128651.1 3, 4 713
NM_016598.2 NP_057682.1 3, 4 713
XM_005265209.1 XP_005265266.1 3, 4 713
XM_005265212.2 XP_005265269.1 3, 4 721
XM_006713189.1 XP_006713252.1 4, 5 815
XM_006713192.2 XP_006713255.1 4, 5 815
XM_006713193.2 XP_006713256.1 4, 5 816
XM_006713194.3 XP_006713257.1 3, 4 696
XM_011533797.1 XP_011532099.1 4, 5 815
XM_011533800.2 XP_011532102.1 4, 5 808
XM_011533801.2 XP_011532103.1 4, 5 815
XM_017006557.1 XP_016862046.1 4, 5 815
XM_017006558.1 XP_016862047.1 5, 6 861
XM_017006559.1 XP_016862048.1 4, 5 910
XM_017006560.1 XP_016862049.1 4, 5 815
XM_017006561.1 XP_016862050.1 3, 4 713
XM_017006562.1 XP_016862051.1 3, 4 713
XM_017006563.1 XP_016862052.1 4, 5 815
XM_017006564.1 XP_016862053.1 4, 5 815
XM_017006565.1 XP_016862054.1 4, 5 796
XM_017006566.1 XP_016862055.1 4, 5 751
XM_017006567.1 XP_016862056.1 4, 5 789
XM_017006568.1 XP_016862057.1 2, 3 222
XM_017006569.1 XP_016862058.1 4, 5 788
XM_017006570.1 XP_016862059.1 3, 4 700
XM_017006571.1 XP_016862060.1 3, 4 709
GenBank mRNA AF247703.1 - 3, 4 653
AF441791.1 - 2, 3 439
AK299782.1 - 1, 2 37
AY605661.1 - 1, 2 133
BC015467.1 - 3, 4 640 3852521

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039014, XM_008766715, XM_008766714, XM_006244188, XM_008766716, and others.
This siRNA also targets the following transcripts in Mouse: NM_026917, BC052464, AB088044, AK018755, AK169197, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein targeting
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0