This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HVCN1 SIRNA Assays ›
Gene Symbol:
HVCN1
Gene Name:
hydrogen voltage gated channel 1
Gene Aliases:
HV1, VSOP
Chromosome Location:
Chr.12: 110648686 - 110704945 on Build GRCh38
UniGene ID:
Hs.211511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 105514

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HVCN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040107.1 NP_001035196.1 5 582
NM_001256413.1 NP_001243342.1 4 542
NM_032369.3 NP_115745.2 5 573
XM_005253948.2 XP_005254005.1 5 660
XM_011538838.1 XP_011537140.1 6 998
XM_011538839.2 XP_011537141.1 6 732
XM_011538840.2 XP_011537142.1 6 764
XM_011538841.2 XP_011537143.1 6 742
XM_011538842.2 XP_011537144.1 6 722
XM_011538844.2 XP_011537146.1 6 737
XM_011538845.1 XP_011537147.1 5 617
XM_011538846.2 XP_011537148.1 6 691
XM_011538847.1 XP_011537149.1 5 592
XM_017020026.1 XP_016875515.1 6 706
XM_017020027.1 XP_016875516.1 5 591
GenBank mRNA AK293543.1 - 4 508
AY359054.1 - 5 601
BC007277.1 - 5 1025 3343102
BC009731.1 - 5 524 3877292
BC032672.2 - 4 1146 5577070
BP369986.1 - 5 455
GU211002.1 - 3 392

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to pH
response to zinc ion
proton transport
ion transmembrane transport
regulation of ion transmembrane transport
sperm-egg recognition
cellular response to zinc ion
cellular response to pH
voltage-gated cation channel activity
voltage-gated proton channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header