This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP6C SIRNA Assays ›
Gene Symbol:
PPP6C
Gene Name:
protein phosphatase 6 catalytic subunit
Gene Aliases:
PP6, PP6C
Chromosome Location:
Chr.9: 125146573 - 125189939 on Build GRCh38
UniGene ID:
Hs.744091
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 104531

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPP6C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123355.1 NP_001116827.1 7, 8 997
NM_001123369.1 NP_001116841.1 5, 6 820
NM_002721.4 NP_002712.1 6, 7 886
XM_011518847.2 XP_011517149.1 6, 7 683
GenBank mRNA AF035158.2 - 6, 7 659
AK295190.1 - 7, 8 862
AK295752.1 - 4, 5 498
AK299087.1 - 5, 6 684
AK312332.1 - 6, 7 718
AK316527.1 - 4, 5 532
BC006990.1 - 6, 7 733 3684339
BT019708.1 - 6, 7 665
KU178271.1 - 6, 7 665
KU178272.1 - 4, 5 457
X92972.2 - 6, 7 686

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein dephosphorylation
COPII vesicle coating
protein serine/threonine phosphatase activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0