This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENDOG SIRNA Assays ›
Gene Symbol:
Endog
Gene Name:
endonuclease G
Gene Aliases:
-
Chromosome Location:
Chr.2: 30171401 - 30174069 on Build GRCm38
UniGene ID:
Mm.474473
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 101900

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Endog
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007931.1 NP_031957.1 2 569
GenBank mRNA AB012108.1 - 2 569
AK144459.1 - 2 601
BC030177.1 - 2 578 5029633
X99394.1 - 2 65

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
DNA metabolic process
DNA catabolic process
apoptotic DNA fragmentation
response to tumor necrosis factor
positive regulation of apoptotic process
response to antibiotic
nucleic acid phosphodiester bond hydrolysis
positive regulation of hydrogen peroxide-mediated programmed cell death
positive regulation of apoptotic DNA fragmentation
nucleic acid binding
nuclease activity
endonuclease activity
deoxyribonuclease activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0