This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CPNE9 SIRNA Assays ›
Gene Symbol:
Cpne9
Gene Name:
copine family member IX
Gene Aliases:
A730016F12Rik, mKIAA4217
Chromosome Location:
Chr.6: 113282266 - 113305571 on Build GRCm38
UniGene ID:
Mm.65597
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100854

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cpne9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170673.3 NP_733773.2 4 346
GenBank mRNA AK042692.1 - 4 347
AK044080.1 - 4 312
AK047431.1 - 1 1038
AK047571.1 - 4 290
AK137422.1 - 4 347
BB639866.1 - 4 347
BC115728.1 - 4 296 40039295

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024982, XM_006237022, XM_008763190, BC091347.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of dendrite extension
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0