This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MDM1 SIRNA Assays ›
Gene Symbol:
Mdm1
Gene Name:
transformed mouse 3T3 cell double minute 1
Gene Aliases:
Arrd2, Mdm-1
Chromosome Location:
Chr.10: 118141537 - 118168999 on Build GRCm38
UniGene ID:
Mm.101191
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100842

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mdm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162904.1 NP_001156376.1 2 278
NM_001162905.1 NP_001156377.1 2 278
NM_010785.2 NP_034915.2 2 278
NM_148922.3 NP_683724.2 2 278
GenBank mRNA AK004789.2 - 2 233
BC052680.1 - 2 228
BC137608.1 - 2 206 8860627
BC144777.1 - 2 206 9053325
EU490427.1 - 2 120
EU490428.1 - 2 120
M20823.1 - 2 141
M20824.1 - 2 141

Back To Top

More Information


Gene Ontology Categories:

Function(s)

retina development in camera-type eye

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0