This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHCHD1 SIRNA Assays ›
Gene Symbol:
Chchd1
Gene Name:
coiled-coil-helix-coiled-coil-helix domain containing 1
Gene Aliases:
1110001O19Rik, 2400010G13Rik, MRP-S37
Chromosome Location:
Chr.14: 20703027 - 20704425 on Build GRCm38
UniGene ID:
Mm.246996
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100719

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chchd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025366.3 NP_079642.1 2 268
GenBank mRNA AK003263.1 - 2 227
AK010311.1 - 2 211
AK011261.1 - 2 226
AK027907.1 - 2 252
AV460497.1 - 2 268
BC049582.1 - 251 6772118
BC052720.1 - 2 247 6818266
BC064310.1 - 2 247
BC092216.1 - 2 235 30609530

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108369, FQ217372, FQ224809, FQ217729, BC168773, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0