This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DGCR6 SIRNA Assays ›
Gene Symbol:
Dgcr6
Gene Name:
DiGeorge syndrome critical region gene 6
Gene Aliases:
-
Chromosome Location:
Chr.16: 18052860 - 18071632 on Build GRCm38
UniGene ID:
Mm.27155
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100417

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dgcr6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289813.1 NP_001276742.1 4 509
NM_001289814.1 NP_001276743.1 4 500
NM_010047.4 NP_034177.1 5 697
GenBank mRNA AF021031.1 - 4 401
AK019346.1 - 5 700
AK160342.1 - 5 687
BC048503.1 - 4 352 6770478
BE945982.1 - 4 409

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0