Assay Details
Gene Symbol: LOC100284750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157645.1 | NP_001151117.1 | 553 | 98 | ||
GenBank mRNA | EU969554.1 | - | 553 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
