Assay Details
Gene Symbol: hda102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111607.1 | NP_001105077.1 | 475 | 79 | ||
GenBank mRNA | AF440228.1 | - | 475 | 79 | ||
AY104306.1 | - | 391 | 79 | |||
BT038157.1 | - | 439 | 79 |
