Assay Details
Gene Symbol: hda110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111625.1 | NP_001105095.1 | 1900 | 82 | ||
GenBank mRNA | AF527611.1 | - | 1900 | 82 | ||
AF527612.1 | - | 1538 | 82 | |||
AF527613.1 | - | 1389 | 82 | |||
AF527614.1 | - | 1195 | 82 | |||
BT033464.1 | - | 1932 | 82 | |||
BT063159.1 | - | 1962 | 82 |
