Assay Details
Gene Symbol: hda110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111625.1 | NP_001105095.1 | 1851 | 98 | ||
GenBank mRNA | AF527611.1 | - | 1851 | 98 | ||
AF527612.1 | - | 1489 | 98 | |||
AF527613.1 | - | 1340 | 98 | |||
AF527614.1 | - | 1146 | 98 | |||
BT033464.1 | - | 1883 | 98 | |||
BT063159.1 | - | 1913 | 98 |
