This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Zm04050061_g1
See other LOC100281965 GE Assays ›
Gene Symbol:
LOC100281965
Gene Name:
3-5 exoribonuclease CSL4
Gene Aliases:
ZEAMMB73_791647
Chromosome Location:
-
Species:
Zea mays
Species Specific ID (Flybase ID):
-
Assay ID Zm04050061_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100281965
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001154882.1 NP_001148354.1 201 68
GenBank mRNA BT066140.1 - 233 68
EU958888.1 - 201 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Zea mays

Panther Classification:

Molecular Function -

RNA metabolism protein exoribonuclease nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0