Assay Details
Gene Symbol: LOC100284313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157208.1 | NP_001150680.1 | 1256 | 104 | ||
GenBank mRNA | BT041677.2 | - | 1252 | 104 | ||
EU967864.1 | - | 1256 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
